Order Entry
Denmark
ContactUsLinkComponent
pH 3.00 at 20 C Citric Acid/ Sodium Chloride/ Hydrochloric Acid CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 500 mL
  ACSDPH103.L5
 :  CPACHEM
New Product
pH 3.00 at 20 C Citric Acid/ Sodium Chloride/ Hydrochloric Acid CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 500 mL
  ACSDPH103.L5
 :  CPACHEM