Order Entry
Denmark
Orders LinkContactUsLinkComponent
Nimorazole

Nimorazole

Nimorazole
Catalog # EHERC15527000
Supplier:   EHRENSTORFER
Nimorazole
Catalog # EHERC15527000
Supplier:  EHRENSTORFER
CAS Number:  
Nimorazole
Nimorazole

Nimorazole

CAS Number:  
Price
2.670,00 kr
Log in for availability
1 * 10 mg
Product Details & Documents
  • SMILES: [O-][N+](=O)c1cncn1CCN2CCOCC2
  • Inchi: InChI=1S/C9H14N4O3/c14-13(15)9-7-10-8-12(9)2-1-11-3-5-16-6-4-11/h7-8H,1-6H2
  • IUPAC: 4-[2-(5-nitroimidazol-1-yl)ethyl]morpholine
Specifications

Specifications

  • Description:
    Nimorazole
  • Cat. No.:
    EHERC15527000
  • Formula:
    C₉H₁₄N₄O₃
  • Storage conditions:
    4 °C
  • Hazard class description:
    GHS07, GHS08
  • GHS signal:
    Danger
  • Synonym(s):
    Morpholine;4-[2-(5-nitro-1H-imidazol-1-yl)ethyl]-;Morpholine;4-[2-(5-nitroimidazol-1-yl)ethyl]- (7CI,8CI);1-(β-Morpholinoethyl)-5-nitroimidazole;4-[2-(5-Nitroimidazol-1-yl)ethyl]morpholine;Acterol;Esclama;K-1900;NSC 107524;Naxofem;Naxogin;Nimorazol;Nimorazole;Nitrimidazine;Nulogyl
  • Molecular weight:
    226,23
  • CAS No.:
    6506-37-2
  • Shipping conditions:
    RT

Specifications