Haloxyfop-R-methyl
Haloxyfop-R-methyl
Catalog # EHERC14062500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COC(=O)[C@@H](C)Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1
- Inchi: InChI=1S/C16H13ClF3NO4/c1-9(15(22)23-2)24-11-3-5-12(6-4-11)25-14-13(17)7-10(8-21-14)16(18,19)20/h3-9H,1-2H3/t9-/m1/s1
- IUPAC: methyl (2R)-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoate
- Packing group: III
Specifications
Specifications
- Description:Haloxyfop-R-methyl
- UN No.:3082
- Cat. No.:EHERC14062500
- Formula:C₁₆H₁₃ClF₃NO₄
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Propanoic acid;2-[4-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]-;methyl ester;(R)-;(R)-Haloxyfop methyl ester;(R)-Haloxyfop-methyl;DE 535;Edge;Eloge;Gallant super;Haloxyfop-P-methyl;Haloxyfop-R-methyl;Zellek Super
- Molecular weight:375,73
- CAS No.:72619-32-0
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



