Ethyl-tert-butyl ether
Ethyl-tert-butyl ether
Katalog # EHERC13321300
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CCOC(C)(C)C
- Inchi: InChI=1S/C6H14O/c1-5-7-6(2,3)4/h5H2,1-4H3
- IUPAC: 2-ethoxy-2-methylpropane
- Packing group: II
Specifikationer
Specifikationer
- Description:Ethyl-tert-butyl ether
- UN No.:1179
- Cat. No.:EHERC13321300
- Formula:C₆H₁₄O
- Storage temperature:20 °C
- Hazard class description:GHS02, GHS07
- GHS signal:Danger
- Synonym(s):Propane;2-ethoxy-2-methyl-;Ether;tert-butyl ethyl (6CI,7CI,8CI);2-Ethoxy-2-methylpropane;1,1-Dimethylethyl ethyl ether;Ethyl 1,1-dimethylethyl ether;Ethyl tert-butyl ether;Ethyl tert-butyl oxide;NSC 1069;tert-Butyl ethyl ether
- Molecular weight:102,17
- CAS No.:637-92-3
- Shipping conditions:RT
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



