Bromochlorophenolblue 95% (dye content)
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent
Synonyms: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromo-6-chlorophenol] S,S-dioxide
- CAS Number:2553-71-1
- Formula:C₁₉H₁₀Br₂Cl₂O₅S
- Molecular Weight:581,06 g/mol
5 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromo-6-chlorophenol] S,S-dioxide
- CAS Number:2553-71-1
- Formula:C₁₉H₁₀Br₂Cl₂O₅S
- Molecular Weight:581,06 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



