Order Entry
Denmark
Orders LinkContactUsLinkComponent
Quercetin ≥95%

Quercetin ≥95%

Toxic
New Product
Supplier:   Apollo Scientific
Quercetin ≥95%
Supplier:  Apollo Scientific
New Product
Certificates
Quercetin ≥95%

2 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

  • Store at –20 °C
Synonyms:  3-5-7-3-4-Pentahydroxyflavone, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 3,5,7,3',4'-Pentahydroxyflavone
  • CAS Number:117-39-5
  • Formula:C₁₅H₁₀O₇
  • MDL Number:MFCD00006828
  • Molecular Weight:302,24 g/mol
  • Storage Temperature:Freezer
  • EINECS:204-187-1
  • UN:2811
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 407,00 kr
    407,00 kr
    1 * 500 mg
       407,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 483,00 kr
    483,00 kr
    1 * 1 g
       483,00 kr

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.