1-Hydroxyurea 98%
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/ reagent.
- Store at 2 to 8 °C
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76,05466 g/mol
- Storage Temperature:Refrigerator
- EINECS:204-821-7
5 Options Available Below
Product Details & Documents
Usage: Organic building block/ reagent.
- Store at 2 to 8 °C
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76,05466 g/mol
- Storage Temperature:Refrigerator
- EINECS:204-821-7
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



