Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-Sorbitol ≥95%

D-Sorbitol ≥95%

New Product
Supplier:   Apollo Scientific
D-Sorbitol ≥95%
Supplier:  Apollo Scientific
New Product
Certificates
D-Sorbitol ≥95%

4 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:C₆H₁₄O₆
  • Molecular Weight:182,17 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 319,00 kr
    319,00 kr
    1 * 50 mg
       319,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 428,00 kr
    428,00 kr
    1 * 100 mg
       428,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 450,00 kr
    450,00 kr
    1 * 200 mg
       450,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 538,00 kr
    538,00 kr
    1 * 500 mg
       538,00 kr

    Specifications:
    New Product
    New Product