Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thymol blue indicator

Thymol blue indicator

Supplier:   Apollo Scientific
Thymol blue indicator
Supplier:  Apollo Scientific
Certificates
Thymol blue indicator

4 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Thymolsulphonphthalein, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:76-61-9
  • Formula:C₂₇H₃₀O₅S
  • MDL Number:MFCD00005869
  • Molecular Weight:466,59 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 165,00 kr
    165,00 kr
    1 * 5 g
       165,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 187,00 kr
    187,00 kr
    1 * 25 g
       187,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 483,00 kr
    483,00 kr
    1 * 100 g
       483,00 kr

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1.620,00 kr
    1.620,00 kr
    1 * 500 g
       1.620,00 kr