Murexide indicator
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent.
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₅N₅O₆·H₃N
- MDL Number:MFCD00012777
- Molecular Weight:284,18572 g/mol
- EINECS:221-266-6
6 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₅N₅O₆·H₃N
- MDL Number:MFCD00012777
- Molecular Weight:284,18572 g/mol
- EINECS:221-266-6
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



