Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

1-Hydroxyoctadecane ≥98%

New Product
Supplier:   Apollo Scientific
1-Hydroxyoctadecane ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
1-Hydroxyoctadecane ≥98%

3 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
  • CAS Number:112-92-5
  • Formula:C₁₈H₃₈O
  • Molecular Weight:270,49 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 472,00 kr
    472,00 kr
    1 * 500 mg
       472,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 681,00 kr
    681,00 kr
    1 * 1 g
       681,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1.630,00 kr
    1.630,00 kr
    1 * 5 g
       1.630,00 kr

    Specifications:
    New Product
    New Product