Order Entry
Denmark
Orders LinkContactUsLinkComponent
L-Hydroxyproline ≥98%

L-Hydroxyproline ≥98%

Bottle
Catalog # APOSBUP18166-500MG
New Product
Supplier:   Apollo Scientific
L-Hydroxyproline ≥98%
Catalog # APOSBUP18166-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  51-35-4
L-Hydroxyproline ≥98%

Bottle

CAS Number:  51-35-4
Price
396,00 kr
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
  • CAS Number:51-35-4
  • Formula:C₅H₉NO₃
  • Molecular Weight:131,13 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications