Order Entry
Denmark
Orders LinkContactUsLinkComponent
DL-alpha-Tocopherol ≥98%

DL-alpha-Tocopherol ≥98%

Bottle
Catalog # APOSBUP17590-200MG
New Product
Supplier:   Apollo Scientific
DL-alpha-Tocopherol ≥98%
Catalog # APOSBUP17590-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  10191-41-0
DL-alpha-Tocopherol ≥98%

Bottle

CAS Number:  10191-41-0
Price
297,00 kr
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
  • CAS Number:10191-41-0
  • Formula:C₂₉H₅₀O₂
  • Molecular Weight:430,71 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.