Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzo[a]pyrene 99%

Benzo[a]pyrene 99%

New Product
Supplier:   Apollo Scientific
Benzo[a]pyrene 99%
Supplier:  Apollo Scientific
New Product
Certificates
Benzo[a]pyrene 99%

4 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Benzo[def]chrysene, BaP
  • CAS Number:50-32-8
  • Formula:C₂₀H₁₂
  • Molecular Weight:252,31 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 308,00 kr
    308,00 kr
    1 * 100 mg
       308,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 615,00 kr
    615,00 kr
    1 * 250 mg
       615,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 2.390,00 kr
    2.390,00 kr
    1 * 1 g
       2.390,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 11.830,00 kr
    11.830,00 kr
    1 * 5 g
       11.830,00 kr

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.