Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

6PPD-Q ≥98%

Corrosive
Environment
Irritant
New Product
Supplier:   Apollo Scientific
6PPD-Q ≥98%
Supplier:  Apollo Scientific
New Product
Certificates

2 Options Available Below

Product Details & Documents

Usage: Biochemical/life science applications

  • CAS Number:2754428-18-5
  • Formula:C₁₈H₂₂N₂O₂
  • MDL Number:MFCD34561806
  • Molecular Weight:298,38 g/mol
  • UN:3077
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 2.870,00 kr
    2.870,00 kr
    1 * 25 mg
       2.870,00 kr

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 4.370,00 kr
    4.370,00 kr
    1 * 50 mg
       4.370,00 kr

    Specifications:
    New Product
    New Product