Phenol Red indicator
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Organic building block/reagent
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354,38 g/mol
- EINECS:205-609-7
6 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:143-74-8
- Formula:C₁₉H₁₄O₅S
- MDL Number:MFCD00003552
- Molecular Weight:354,38 g/mol
- EINECS:205-609-7
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



