Order Entry
Denmark
Orders LinkContactUsLinkComponent
Jupiter HPLC Columns, Proteo
Jupiter HPLC Columns, Proteo
New Product
Supplier:   PHENOMENEX
Jupiter HPLC Columns, Proteo
Supplier:  PHENOMENEX
New Product
Certificates
Jupiter HPLC Columns, Proteo
15 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • PHEC00B-4396-AC
  • PHEC00F-4396-AC
  • PHEC00F-4396-AF
  • PHEC00B-4396-A0
  • PHEC00B-4396-B0
  • PHEC00F-4396-B0
  • PHEC00G-4396-B0
  • PHEC00F-4396-E0
  • PHEC00G-4396-E0
  • PHEC00G-4396-N0
  • PHEC00G-4396-P0-AX
  • PHEC00D-4396-U0-AX
  • PHEC00F-4396-U0-AX
  • PHEC00G-4396-U0-AX
  • PHEC00G-4397-V0-AX
  • Carbon load:
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • 15%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • pH range:
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • 1,5 - 10,0
  • Surface area:
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
  • 475 m²/g
Specifications
Product Family Options
Columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Capillary
    Proteo
    Stainless steel, glass coated
    300 Å
    4 µm
    0,30 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 12.042,00 kr
    12.042,00 kr
    1 * 1 Items
       12.042,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00B-4396-AC
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Capillary
    Proteo
    Stainless steel, glass coated
    300 Å
    4 µm
    0,30 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 11.579,00 kr
    11.579,00 kr
    1 * 1 Items
       11.579,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00F-4396-AC
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Capillary
    Proteo
    Stainless steel, glass coated
    300 Å
    4 µm
    0,50 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 11.579,00 kr
    11.579,00 kr
    1 * 1 Items
       11.579,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00F-4396-AF
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Microbore
    Proteo
    Stainless steel
    300 Å
    4 µm
    1,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.025,00 kr
    9.025,00 kr
    1 * 1 Items
       9.025,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00B-4396-A0
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Microbore
    Proteo
    Stainless steel
    300 Å
    4 µm
    2,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.025,00 kr
    9.025,00 kr
    1 * 1 Items
       9.025,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00B-4396-B0
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Microbore
    Proteo
    Stainless steel
    300 Å
    4 µm
    2,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10.261,00 kr
    10.261,00 kr
    1 * 1 Items
       10.261,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00F-4396-B0
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Microbore
    Proteo
    Stainless steel
    300 Å
    4 µm
    2,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10.941,00 kr
    10.941,00 kr
    1 * 1 Items
       10.941,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00G-4396-B0
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Analytical
    Proteo
    Stainless steel
    300 Å
    4 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10.434,00 kr
    10.434,00 kr
    1 * 1 Items
       10.434,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00F-4396-E0
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Analytical
    Proteo
    Stainless steel
    300 Å
    4 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10.680,00 kr
    10.680,00 kr
    1 * 1 Items
       10.680,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00G-4396-E0
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Semi-preparative
    Proteo
    Stainless steel
    300 Å
    4 µm
    10,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 23.533,00 kr
    23.533,00 kr
    1 * 1 Items
       23.533,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00G-4396-N0
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
AXIA™ packed columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Preparative
    Proteo
    Stainless steel
    300 Å
    4 µm
    21,20 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 49.249,00 kr
    49.249,00 kr
    1 * 1 Items
       49.249,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00G-4396-P0-AX
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Preparative
    Proteo
    Stainless steel
    300 Å
    4 µm
    30,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 63.709,00 kr
    63.709,00 kr
    1 * 1 Items
       63.709,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00D-4396-U0-AX
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Preparative
    Proteo
    Stainless steel
    300 Å
    4 µm
    30,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 96.992,00 kr
    96.992,00 kr
    1 * 1 Items
       96.992,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00F-4396-U0-AX
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Preparative
    Proteo
    Stainless steel
    300 Å
    4 µm
    30,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 144.880,00 kr
    144.880,00 kr
    1 * 1 Items
       144.880,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00G-4396-U0-AX
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g
    New Product
  • Preparative
    Proteo
    Stainless steel
    300 Å
    10 µm
    50,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 159.563,00 kr
    159.563,00 kr
    1 * 1 Items
       159.563,00 kr
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    PHEC00G-4397-V0-AX
    Carbon load
    15%
    Endcapped
    Yes
    pH range
    1,5 - 10,0
    Surface area
    475 m²/g