Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tip-off Manifolds
Tip-off Manifolds
Complete Tip-Off Manifold, outer diameter 4 mm
Catalog # WILM552-4
New Product
Supplier:   SP Industries
Tip-off Manifolds
Catalog # WILM552-4
Supplier:  SP Industries
New Product
Tip-off Manifolds
Complete Tip-Off Manifold, outer diameter 4 mm
Price
1.730,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The Tip-Off Manifold connects to an NMR tube by a threaded aluminum bushing which is isolated from the vacuum by a PTFE high-vacuum rotary valve with Viton O-rings. Rotating the valve will open and close the tube to the vacuum line.
  • Highly resistant to chemicals
    Easy operation
Specifications
  • Description:
    Complete Tip-Off Manifold, outer diameter 4 mm
  • Seq:
    2