Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Serine solution ≥98% NMDA receptor co-agonist

D-(-)-Serine solution ≥98% NMDA receptor co-agonist

Catalog # ABCAAB146671-100
Supplier:   Abcam
D-(-)-Serine solution ≥98% NMDA receptor co-agonist
Catalog # ABCAAB146671-100
Supplier:  Abcam
CAS Number:  312-84-5
D-(-)-Serine solution ≥98% NMDA receptor co-agonist
CAS Number:  312-84-5
Price
549,00 kr
Log in for availability
1 * 100 mmol
Product Details & Documents

Agonist at the nMDA glycine binding site and the inhibitory post-synaptic glycine receptor.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (R)-(-)-Serine, H-D-Ser-OH
  • CAS Number:312-84-5
  • Molecular Weight:105,09 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% D-Serine
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.