Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Serine ≥99% Fluorogenic (MeOSuc-AAPV-AFC)

D-(-)-Serine ≥99% Fluorogenic (MeOSuc-AAPV-AFC)

Catalog # ABCAAB142178-5
Supplier:   Abcam
D-(-)-Serine ≥99% Fluorogenic (MeOSuc-AAPV-AFC)
Catalog # ABCAAB142178-5
Supplier:  Abcam
CAS Number:  312-84-5
D-(-)-Serine ≥99% Fluorogenic (MeOSuc-AAPV-AFC)
CAS Number:  312-84-5
Price
1.690,00 kr
Log in for availability
1 * 5 mg
Product Details & Documents

Highly sensitive coumarin-fluorogenic substrate for neutrophil elastase (Km =130 μM for leukocyte elastase). Compatible with in vitroassays with purified enzyme, on live cells, or in conditioned medium. The excitation wavelength is 380 nm and the emission wavelength is 500 nm.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  (R)-(-)-Serine, H-D-Ser-OH
  • CAS Number:312-84-5
  • Molecular Weight:681,67 g/mol
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.