Order Entry
Denmark
Orders LinkContactUsLinkComponent
AEC Substrate Buffer, Abcam

AEC Substrate Buffer, Abcam

AEC Substrate Buffer
Catalog # ABCAAB286857-50
Supplier:   Abcam
AEC Substrate Buffer, Abcam
Catalog # ABCAAB286857-50
Supplier:  Abcam
CAS Number:  
AEC Substrate Buffer, Abcam

AEC Substrate Buffer

CAS Number:  
Price
1.220,00 kr
Log in for availability
1 * 50 mL
Product Details & Documents

This product is manufactured by BioVision, an Abcam company and was previously called 2123 AEC Substrate Buffer.

  • Constituents: 2.63% Sodium acetate, 0.3% Hydrogen peroxide

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    AEC Substrate Buffer
  • Cat. No.:
    ABCAAB286857-50
  • Storage temperature:
    Store at +4°C.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.