Order Entry
Denmark
Orders LinkContactUsLinkComponent
Potassium hydroxide solution 2N

Potassium hydroxide solution 2N

Supplier:   G-Biosciences
Potassium hydroxide solution 2N
Supplier:  G-Biosciences
Certificates
Potassium hydroxide solution 2N

2 Options Available Below

Product Details & Documents

A strong, standardized alkaline solution commonly used in biochemistry for pH adjustment, tissue digestion, protein hydrolysis, and cell lysis.

  • Synonym: Caustic potash CAS number: 1310-58-3 Molecular formula: KOH Molecular weight: 56.11

Its strong base properties make it essential in various analytical and preparative procedures where precise control of alkalinity is required.

Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol

Documents

Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1.290,00 kr
    1.290,00 kr
    1 * 500 mL
       1.290,00 kr

  • Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 4.180,00 kr
    4.180,00 kr
    1 * 1 L
       4.180,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.