Millipore® Immobilon®-E NOW Bundle Hydrophobic PVDF Transfer Membrane
Catalog # MILFNWKITIE
Supplier: Merck
New Product
Specifications
- Description:Immobilon®-E NOW bundle PVDF transfer membrane
- Dimensions:85×10000 mm
- Environmentally Preferable:
Specifications
About this item
The first PVDF transfer membrane for western blotting that wets out in transfer buffer or water. For chemiluminescence or chromogenic detection.
Western Blotting
Immunoblotting (Western)
Dot Blot
White/plain filter
Electrostatic, hydrophobic
Easily wetted with water
Immobilon-E is the first PVDF membrane for Western blotting that can be wetted with with aqueous transfer buffer. This eliminates the alcohol pre-wet step required with other PVDF membranes.
45 µm pore size recommended for most Western blotting applications, especially proteins greater than 20 kDa.
Compatible stains: Coomassie brilliant blue, Amido black, India ink, Ponceau-S red, Colloidal gold, Transillumination and Sypro® ruby.