Order Entry
Denmark
ContactUsLinkComponent
 
UV/VIS Spectrophotometers
New Product
 
undefined
UV/VIS Spectrophotometers
New Product

 

  • Catalog No:
  • 634-1244
  • 634-1243
  • Wavelength accuracy:
  • ±0,3 nm
  • ±0,3 nm
  • Wavelength range:
  • 190 - 1100 nm
  • 190 - 1100 nm
  • Light source:
  • Double-beam
  • Double-beam
  • Spectral bandwidth:
  • 1,5 nm
  • 1,5 nm
  • W×D×H:
  • 572×500×285 mm
  • 572×500×285 mm
  • Weight:
  • 31 kg
  • 31 kg

 

Frequently Bought Together