Mouse Thyroid Peroxidase/TPO Antibody Pair - BSA and Azide free
Catalog # ABCAAB309253-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Thyroid Peroxidase/TPO antibody pair - BSA and Azide free
- Storage temperature:+4 ℃
- Tests per kit:10×96 tests
Specifications
About this item
Mouse Thyroid Peroxidase/TPO Antibody Pair - BSA and Azide free.