Order Entry
Denmark
ContactUsLinkComponent
Human C3a anaphylatoxin Antibody Pair - BSA and Azide free
Human C3a anaphylatoxin Antibody Pair - BSA and Azide free
Catalog # ABCAAB316993-10X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Human C3a anaphylatoxin Antibody Pair - BSA and Azide free
Catalog # ABCAAB316993-10X96
Supplier:  Abcam
New Product
CAS Number:  
About this item
Human C3a anaphylatoxin Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human C3a anaphylatoxin.