Phospho-p53 (S15) and Total p53 ELISA Kit
Catalog # ABCAAB279878-1X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:p53
- Description:Phospho-p53 (S15) and Total p53 ELISA kit
- Sample type:Cell lysate
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Phospho-p53 (S15) and Total p53 ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phospho-p53 (S15) and Total p53 in Human in Cell Lysate samples.