Mouse MMP12 ELISA Kit
Catalog # ABCAAB246540-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse MMP12 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse MMP12 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse MMP12 with a sensitivity of 32,858 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.