Human Glucose 6 Phosphate Dehydrogenase Antibody Pair - BSA and Azide Free
Catalog # ABCAAB303688-10X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Glucose 6 Phosphate Dehydrogenase
- Description:Human Glucose 6 Phosphate Dehydrogenase antibody pair - BSA and azide free
- Detection range:0,08 - 10 ng/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human Glucose 6 Phosphate Dehydrogenase antibody pair - BSA and Azide free.