Order Entry
Denmark
ContactUsLinkComponent
Rabbit IL-2 ELISA Kit
Rabbit IL-2 ELISA Kit
Catalog # ABCAAB273239-1X96
Supplier:  Abcam
CAS Number:  
undefined
Rabbit IL-2 ELISA Kit
Catalog # ABCAAB273239-1X96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Rabbit IL-2 ELISA kit
Specifications
About this item
Rabbit IL-2 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rabbit IL-2 in Rabbit in plasma, cell culture supernatant, serum samples.