Human/Mouse Hyaluronan synthase 2 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB290699-960
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Hyaluronan synthase 2
- Description:Human/Mouse Hyaluronan synthase 2 antibody pair - BSA and azide free
- Detection range:1,56 - 100 ng/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human/Mouse Hyaluronan synthase 2 antibody pair - BSA and Azide free.