Order Entry
Denmark
ContactUsLinkComponent
 
Human/Mouse c-Fos Antibody Pair - BSA and Azide Free
Catalog # ABCAAB267732-10X96
Supplier:  Abcam
CAS Number:  
undefined
Human/Mouse c-Fos Antibody Pair - BSA and Azide Free
Catalog # ABCAAB267732-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    c-Fos
  • Description:
    Human/Mouse c-Fos antibody pair - BSA and azide free
  • Detection range:
    54,6875 - 3500 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

Specifications

About this item

Human/Mouse c-Fos antibody pair - BSA and Azide free.