Human Prolactin Receptor/PRL-R Antibody Pair - BSA and Azide free
Catalog # ABCAAB316820-10X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Description:Human Prolactin Receptor/PRL-R antibody pair - BSA and Azide free
- Storage temperature:+4 ℃
- Tests per kit:10×96 tests
Specifications
About this item
Human Prolactin Receptor/PRL-R Antibody Pair - BSA and Azide free.