SPON2 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB302767-960
Supplier: Abcam
CAS Number:
Specifications
- Description:SPON2 Antibody Pair - BSA and Azide free
- Tests per kit:10×96 tests
Specifications
About this item
SPON2 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of SPON2,