Mouse/Rat FRZB Antibody Pair - BSA and Azide Free
Catalog # ABCAAB253827-10X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:FRZB
- Description:Mouse/Rat FRZB antibody pair - BSA and azide free
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Mouse/Rat FRZB antibody pair - BSA and Azide free.