Order Entry
Denmark
ContactUsLinkComponent
 
Mouse C Reactive Protein Antibody Pair - BSA and Azide Free
  ABCAAB241785-10X96
 :  Abcam
 :  
undefined
Mouse C Reactive Protein Antibody Pair - BSA and Azide Free
  ABCAAB241785-10X96
 :  Abcam
 :  

 

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    C Reactive Protein
  • Description:
    Mouse C Reactive Protein antibody pair - BSA and azide free
  • Detection range:
    125 - 8000 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

 

 

Mouse C Reactive Protein antibody pair - BSA and Azide free.