Order Entry
Denmark
ContactUsLinkComponent
Mouse Fas Ligand ELISA Kit (FasL)
Mouse Fas Ligand ELISA Kit (FasL)
Catalog # ABCAAB270894-1X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse Fas Ligand ELISA Kit (FasL)
Catalog # ABCAAB270894-1X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Fas Ligand
  • Description:
    Mouse Fas Ligand ELISA kit (FasL)
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    18,75 - 1200 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    5,7 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse Fas Ligand ELISA kit (FasL) is a single-wash 90-min Simplestep used to quantify Mouse Fas Ligand (FasL) with a sensitivity of 5.7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.