Human Carbonic Anhydrase 9/CA9 Antibody Pair - BSA and Azide free
ABCAAB253844-10X96
: Abcam
:
- Description:Human Carbonic Anhydrase 9/CA9 Antibody Pair - BSA and Azide free
- Tests per kit:10×96 tests
Human Carbonic Anhydrase 9/CA9 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Carbonic Anhydrase 9/CA9.