Order Entry
Denmark
ContactUsLinkComponent
Phosphotyrosine JAK2 ELISA Kit
Phosphotyrosine JAK2 ELISA Kit
Catalog # ABCAAB279839-1X96
Supplier:  Abcam
CAS Number:  
undefined
Phosphotyrosine JAK2 ELISA Kit
Catalog # ABCAAB279839-1X96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Phosphotyrosine JAK2 ELISA kit
Specifications
About this item
Phosphotyrosine JAK2 ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phosphotyrosine JAK2 in Human in cell Lysate samples.