Mouse DKK1 ELISA Kit (Dickkopf-1)
Catalog # ABCAAB113347-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:DKK1
- Description:Mouse DKK1 ELISA kit (Dickkopf-1)
- Sample type:Plasma, cell culture supernatant
- Detection method:Colorimetric
- Detection range:8,23 - 6000 pg/ml
- Storage temperature:4 °C
- Sensitivity:25 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse DKK1 ELISA kit (Dickkopf-1) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse DKK1 (Dickkopf-1) in Mouse in plasma, cell culture supernatant samples.