Mouse LIX ELISA Kit (CXCL5)
Catalog # ABCAAB264611-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse LIX ELISA Kit (CXCL5)
- Tests per kit:96 tests
Specifications
About this item
Mouse LIX ELISA Kit (CXCL5) is a single-wash 90-min Simplestep used to quantify Mouse LIX (CXCL5) with a sensitivity of 1,14 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.