Human Fibrinogen gamma chain Antibody Pair - BSA and Azide Free
Catalog # ABCAAB253774-10X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Fibrinogen gamma chain
- Description:Human Fibrinogen gamma chain antibody pair - BSA and azide free
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human Fibrinogen gamma chain antibody pair - BSA and Azide free.