Human/Mouse LRRK2 Antibody Pair - BSA and Azide free
ABCAAB307747-10X96
: Abcam
:
- Description:Human/Mouse LRRK2 antibody pair - BSA and Azide free
- Storage temperature:+4 ℃
- Tests per kit:10×96 tests
Human/Mouse LRRK2 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human/Mouse LRRK2.