Mouse Flt3 ligand Antibody Pair - BSA and Azide free (Flt3L)
Catalog # ABCAAB253443-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Flt3 ligand antibody pair - BSA and Azide free (Flt3L)
- Storage temperature:+4 ℃
- Tests per kit:10×96 tests
Specifications
About this item
Mouse Flt3 ligand Antibody Pair - BSA and Azide free. (Flt3L)