Order Entry
Denmark
ContactUsLinkComponent
Mouse GC-C ELISA Kit
Mouse GC-C ELISA Kit
Catalog # ABCAAB246544-1X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse GC-C ELISA Kit
Catalog # ABCAAB246544-1X96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse GC-C ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse GC-C ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse GC-C with a sensitivity of 11 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.