Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sample Diluent NBS

Sample Diluent NBS

Sample diluent NBS, 20 ml
Catalog # ABCAAB221827-20
Supplier:   Abcam
Sample Diluent NBS
Catalog # ABCAAB221827-20
Supplier:  Abcam
CAS Number:  
Sample Diluent NBS
Sample Diluent NBS

Sample diluent NBS, 20 ml

CAS Number:  
Price
588,00 kr
Log in for availability
1 * 20 mL
Product Details & Documents

Sample Diluent NBS (ab221827) is part of the reagents, controls and accessories range. High-quality biological reagents and tools including antibodies, proteins, assays, cell lines and lysates are available.

Caution: RUO

Specifications

Specifications

  • Description:
    Sample diluent NBS
  • Size:
    20 ml
  • Cat. No.:
    ABCAAB221827-20
  • Storage temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.