Human Arg2 Antibody Pair - BSA and Azide free
Catalog # ABCAAB253756-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Arg2 Antibody Pair - BSA and Azide free
Specifications
About this item
Human Arg2 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Arg2.