Human PAI1 + tPA Complex Antibody Pair - BSA and Azide free
Catalog # ABCAAB253833-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human PAI1 + tPA Complex antibody pair - BSA and Azide free
- Storage temperature:+4 ℃
- Tests per kit:10×96 tests
Specifications
About this item
Human PAI1 + tPA Complex Antibody Pair - BSA and Azide free.