Order Entry
Denmark
Orders LinkContactUsLinkComponent
Monkey IgM ELISA kit

Monkey IgM ELISA kit

Monkey IgM ELISA kit, 96 tests
Catalog # ABCAAB211646-96
Supplier:   Abcam
Monkey IgM ELISA kit
Catalog # ABCAAB211646-96
Supplier:  Abcam
CAS Number:  
Monkey IgM ELISA kit
Monkey IgM ELISA kit

Monkey IgM ELISA kit, 96 tests

CAS Number:  
Price
6.040,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Monkey IgM ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Monkey IgM in Monkey in other biological fluids samples.

Specifications

Specifications

  • Description:
    Monkey IgM ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Monkey
  • Sample type:
    Other biological fluids
  • Detection range:
    12,5 - 400 ng/ml
  • Storage temperature:
    4 ℃
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 40 min
  • Cat. No.:
    ABCAAB211646-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    4,265 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    IgM
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.