Order Entry
Denmark
ContactUsLinkComponent
Bag sealer, electronic
Bag sealer, electronic
Catalog # SEVEFS3604
undefined
Bag sealer, electronic
Catalog # SEVEFS3604

Specifications

  • Description:
    Electronic bag sealer, FS 3604, with automatic recovery, output 160 W
  • Pk:
    1

Specifications

About this item

For vacuum sealing bags.

  • Air suction function
  • Sealing wire and cut-off wire
  • Viewing window
  • Pilot lights for both vacuum control and sealing function
  • Roll of sealing film (width 28,5 cm) included
  • Cable storage

W×D×H: 355×142×100 mm